Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntenin 1 Antibody (K10P3D5*H3), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250287C
Description
Syntenin 1 Monoclonal specifically detects Syntenin 1 in Human samples. It is validated for Western Blot.Specifications
Syntenin 1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SDCBP | |
Peptide - HIIKRMAPS | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
K10P3D5*H3 | |
Western Blot | |
MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1 | |
Mouse | |
Protein A purified | |
RUO | |
6386 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction