Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntenin 1 Antibody (OTI2H6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274418IR
Description
Syntenin 1 Monoclonal specifically detects Syntenin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Syntenin 1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SDCBP | |
Full length recombinant protein of human Syntenin 1 (NP_005616) produced in HEK293T cells. | |
0.1 mL | |
6386.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2H6 | |
Western Blot, Immunohistochemistry | |
MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction