Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntenin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582846 Supplier Invitrogen™ Supplier No. PA582846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANVAVVSGAP LQGQLVARPS SINYMVAPVT GNDVGIRRAE IKQGIREVIL CKDQDGKIGL RLKSI Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 89%.

The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntenin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDCBP
Gene Accession No. O00560
Gene Alias MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349)
Gene Symbols SDCBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDCBP. Recombinant protein control fragment (Product #RP-93800).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6386
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.