Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Syntenin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531060
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (85%), Rat (86%), Pig (84%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SDCBP2 contains 2 PDZ (DHR) domains. The function of SDCBP2 remains unknown.
Specifications
Syntenin 2 | |
Polyclonal | |
Unconjugated | |
SDCBP2 | |
SDCBP2; Similar to TACIP18; SITAC; SITAC18; ST-2; syndecan binding protein (syntenin) 2; syndecan binding protein 2; syndecan-binding protein 2; Syntenin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27111 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H190 | |
SDCBP2 | |
Recombinant protein encompassing a sequence within the center region of human Syntenin 2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction