Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Synthegel™ 3D hiPSC Grow Mix, 1 mg, Lyophilized Powder
Shop All Corning Products
Click to view available options
Quantity:
1 mg
Description
- Matrix rigidity is tunable by altering the peptide concentration
- Purified synthetic peptide in neutral pH
- Cells are encapsulated in physiological mimicking conditions
- Fast hydrogel formation (5 to 30 minutes)
- Sol-gel transfer reversible by shearing force
- Works with standard cell culture methodologies, growth factors and media
- Allows growth and passaging of hiPSCs in 3D format
- The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
- Allows 3D embedded and suspension hiPSC cultures

Specifications
Specifications
Content And Storage | Store at 2 to 8°C |
Description | 1 mg hiPSC Growth Mix Supplement |
Culture Type | 3D Cell Culture |
Form | Lyophilized |
Product Type | Extracellular Matrix |
Sterility | Non-sterile |
For Use With | hiPSC 3D Culture Matrix Kits |
Quantity | 1 mg |
Source | Chemically Defined |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction