Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthetic Seawater, ASTM D 1141 Substitute Ocean Water without Heavy Metals, Ricca Chemical
SDP

Catalog No. 83631 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
4 L
20 L
This item is not returnable. View return policy

Catalog No. 8363-1

Supplier: Ricca Chemical Company 83631

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5, 7647-14-5, 7791-18-6, 7757-82-6, 10035-04-8, 7447-40-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name water
SMILES O

Specifications

CAS Min % 94.0, 2.26, 0.78, 0.38, 0.11, 0.06
CAS Max % 98.0, 2.35, 0.81, 0.39, 0.11, 0.06
Grade Laboratory
Packaging Natural Poly Bottle
pH 8.22
Quantity 4 L
Chemical Name or Material Synthetic Sea Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.