Learn More
Fisherbrand™ Synthetic-Tipped Applicators
Catalog No. 22363166 Shop All Fisherbrand Products
Change view
Click to view available options
Handle Material:
Plastic
Wood
Quantity:
1/envelope
2/envelope
Individual
Sterility:
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Handle Material Quantity Sterility
22-363-166 Wood Individual Sterile
22-363-164 Plastic 2/envelope Sterile
22-363-170 Plastic 1/envelope Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-363-166
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Polyester-tipped applicators on wooden or plastic handles
  • Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
  • Sterile and non-sterile products available (see product specifications)
  • Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
  • Swabs are not certified DNA, DNase or RNase free
  • Not suitable for Forensic use or Human Genetic testing
Specifications
Handle Material Wood
Quantity Individual
Tip Material Polyester
For Use With (Application) Specimen collection and transport system
Product Type Applicator
Sterility Sterile