Learn More
Synthware™ Claisen Adapter with 45° Side Arm
GREENER_CHOICESDP
Catalog No. p-6613059
Click to view available options
Standard Taper Joint No.:
14/20
24/40
Claisen adapter with an additional standard taper outer joint at a 45 deg. angle to the vertical outer joint
  • Three-inlet type Claisen adapter for multiple entries into a single neck flask
  • One side joint is at a 45° angle
  • All joints are the same size
Specifications
Material Borosilicate Glass (3.3)
Product Type Claisen Adapter
Standard Taper Joint No. 14/20
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.