Learn More
Synthware™ Connecting Adapter
Click to view available options
Standard Taper Joint No.:
14/20
24/40
Description
Synthware connecting adapter is a 3-way adapter used for connecting three vessels. It has one top outer joint and two lower inner joints, all being the same size.
Specifications
Specifications
For Use With (Application) | Organic Synthesis |
Material | Borosilicate Glass (3.3) |
Product Type | Glass Joint Adapter |
Quantity | 1 |
Standard Taper Joint No. | 24/40 |