Learn More
Synthware™ Connecting Adapter
GREENER_CHOICESDP
Catalog No. p-6613055 Shop All Kemtech Products
Click to view available options
Standard Taper Joint No.:
14/20
24/40
Catalog No. 31-500-689
Supplier: Synthware™ A101420
Only null left
Add to Cart
Add to Cart
Three-arm glass joint adapters for the assembly of three vessels
Synthware connecting adapter is a 3-way adapter used for connecting three vessels. It has one top outer joint and two lower inner joints, all being the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Glass Joint Adapter
Quantity 1
Standard Taper Joint No. 14/20
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.