Learn More
Synthware™ Drying Tube Adapters, Straight
GREENER_CHOICESDP
Catalog No. 31500650
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
With top standard taper outer joint and lower inner joint of same size
Synthware straight drying tube. With top outer and lower inner standard taper joints of the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Drying Tube Adapter
Quantity 1
Standard Taper Joint No. 19/22
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.