Learn More
Synthware™ Enlarging Adapters
Click to view available options
Standard Taper Joint No.:
19/22 Top, 14/20 Bottom
Top: 24/40, Bottom: 14/20
Top: 24/40, Bottom: 19/22
Top: 29/42, Bottom: 14/20
Top: 29/42, Bottom: 24/40
Top: 34/45, Bottom: 24/40
Top: 40/50, Bottom: 24/40
Top: 45/50, Bottom: 29/42
Description
Synthware enlarging adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is smaller in size than the top outer joint.
Specifications
Specifications
For Use With (Application) | Organic Synthesis |
Material | Borosilicate Glass (3.3) |
Product Type | Enlarging Adapter |
Quantity | 1 |
Standard Taper Joint No. | Top: 45/50, Bottom: 29/42 |