Learn More
Synthware™ Flushig Adapters
GREENER_CHOICESDP
Catalog No. p-6613069 Shop All Kemtech Products
Click to view available options
Standard Taper Joint No.:
14/20
24/40
Catalog No. 31-500-585
Supplier: Synthware™ A761420
Only null left
Add to Cart
Add to Cart
For evacuation and transfer of product into another vessel with same size standard taper joints
T-bore PTFE stopcock has a 2mm bore
Specifications
Includes 2mm T-bore PTFE stopcock
Material PTFE
Product Type Flushig Adapter
Standard Taper Joint No. 14/20
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.