Learn More
Synthware™ Flushing Adapters with PTFE Stopcock
GREENER_CHOICESDP
Catalog No. p-6613101
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
Allows systems to be flushed with an inert gas while sampling directly through the bore of the T-bore stopcock
Synthware flushing adapter is used to flush and fill attached vessels with an inert gas to provide protective storage.
Specifications
For Use With (Application) Organic Synthesis
Item Description Flushing Adapter with PTFE Stopcock
Material Borosilicate Glass (3.3)
Product Type Flushing Adapter
Quantity 1
Standard Taper Joint No. 14/20
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.