Learn More
Synthware™ Inlet Adapters
GREENER_CHOICESDP
Catalog No. p-6582269
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
For use with gas inlet tubes or plain stem thermometers
Supplied with one holed rubber adapter
Specifications
Material Borosilicate Glass
Product Type Inlet Adapter
Standard Taper Joint No. 14/20
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.