Description
- Features top standard taper outer joint and and lower drip tip joint that are of same size
Specifications
Specifications
| For Use With (Equipment) | Round Bottom Flask |
| For Use With (Application) | Organic Synthesis |
| Material | Glass |
| Type | West Condenser |
| Standard Taper Joint No | 19/22 |
| Length (Metric) Jacket | 200 mm |