Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Syntri Safeguard Tissue Processing Reagents

Catalog No. 9517ss Shop All Epredia Products
Click to view available options
Product Type:
10% Neutral Buffered Formalin
Clear-Rite 3
Dehydrant 100%
Flex 100
Formal-Fixx
Isopropyl Alcohol
Xylene
Xylene Substitute
This item is not returnable. View return policy

Catalog No. 9517SS

Supplier: Epredia™ 9517SS

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tissue processing reagents that feature RFID technology for reagent management on the Epredia™ Revos™ Tissue Processor.

The Syntri Safeguard Reagents come in 5 liter bottles for tissue processing with RFID tags embedded in the labels. When partnered with the Epredia™ Revos™ Tissue Processor, Syntri Safeguard Reagent usage is tracked and logged on the instrument. The technology remembers every bottle loaded, documents every protocol executed and maintenance performed. Helping to reduce downtime and facilitates proper use of reagents.

Specifications

Product Type Isopropyl Alcohol
Quantity 4 x 5L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.