Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntrophin alpha 1 Antibody (1E1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222587AF532

Catalog No. NP22287A32


Only null left
Add to Cart

Description

Description

Syntrophin alpha 1 Monoclonal specifically detects Syntrophin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Syntrophin alpha 1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
SNTA1
Recombinant human SNTA1 (1-505aa) purified from E. coli
0.1 mL
Primary
Human
IgG2b κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
10
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin
Mouse
Protein A purified
Cytoskeleton Markers, Neuroscience, Vision
6640.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.