Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synuclein alpha/beta Antibody (3B6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122957C
Description
Synuclein alpha/beta Monoclonal specifically detects Synuclein alpha/beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Synuclein alpha/beta | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SNCA | |
This Synuclein alpha/beta Antibody (3B6) was developed against recombinant human Synuclein alpha/beta (119-140aa) purified from E. coli. | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
6622 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
3B6 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
beta-synuclein, I(2)-synuclein, I+/--synuclein, NACP, non A-beta component of AD amyloid, PARK1, PD1, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human alpha-synuclein (alpha-synuclein 119-140aa) mAb, clone 3B6, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human alpha-synuclein protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction