Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synuclein alpha/beta Recombinant Rabbit Monoclonal Antibody (ST05-21)
GREENER_CHOICE

Catalog No. PIMA532176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532176 Supplier Invitrogen™ Supplier No. MA532176
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synuclein alpha/beta
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone ST05-21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Snca
Gene Accession No. O55042, P37377, P37840, Q16143, Q63754, Q91ZZ3
Gene Alias AI838531; alphaSYN; alpha-synuclein; betaSYN; Beta-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Phosphoneuroprotein 14; PNP 14; SNCA; Sncb; Syn; synuclein alpha; synuclein alpha-140; synuclein beta; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor); synuclein, beta
Gene Symbols Snca, SNCB
Host Species Rabbit
Immunogen Synthetic peptide within Human alpha+beta Synuclein aa 2-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104069, 113893, 20617, 29219, 6620, 6622
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.