Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein beta Polyclonal Antibody, Abbomax

Catalog No. 502203666
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3666 Supplier Abbomax Inc Supplier No. 5002774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human Normal Cerebral Cortex Cellular location: Cytoplasm, cytosol, Membrane, Nucleus, Cell junction,.

Synuclein beta Polyclonal antibody specifically detects Synuclein beta in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Synuclein beta
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene SNCB
Gene Accession No. Q16143, Q63754
Gene Alias AI838531; betaSYN; beta-synuclein; Phosphoneuroprotein 14; PNP 14; Sncb; synuclein beta; synuclein, beta
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of Synuclein beta
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113893, 6620
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.