Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α-Synuclein, Mouse, Unlabeled, Clone: SOY1,

Catalog No. MABN1818 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1818 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1818 Supplier MilliporeSigma™ Supplier No. MABN1818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha-synuclein (UniProt P37840; also known as NACP, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, Synuclein alpha-140) is encoded by the SNCA (also known as NACP, PARK1, PARK4) gene (Gene ID 6622) in human. Pathological aggregates are common features of many neurodegenerative diseases, such as tau neurofibrillary tangles (NFTs) in Alzheimer's disease (AD) and frontotemporal degeneration, and α-synuclein (α-syn or αS) Lewy bodies (LBs) in Parkinson's disease (PD) and dementia with LBs (DLB). Alpha-synuclein is a phospholipid-binding protein concentrated in presynaptic terminals where it promotes SNARE complex formation and modulates synaptic functions. Alpha-synuclein is the major component of pathologic inclusions that characterize PD, DLB, and multiple system atrophy (MSA). Research shows that αS exists not only as unfolded monomers, but in large part also as multimers, principally as ∽60kDa tetramers composed of four N-acetylated αS, that assume α-helical conformation and resist aggregation. PD-causing αS missense mutations are found to shift cellular αS from tetramers/multimers to monomers, indicating that decreased α-helical tetramers and increased unfolded monomers initiate pathogenesis. In addition, both casein kinase-1 (CK-1) and CK-2 can catalyze the phosphorylation of αS on Ser129, and Ser129-phosphorylated αS is found in αS inclusions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Synuclein
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SOY1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SNCA, NACP, PARK1, PAR
Gene Accession No. P37840
Host Species Mouse
Immunogen Purified human erythrocyte alpha-synuclein.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000336
Test Specificity Clone SOY1 detected both wild-type alpha-synuclein and fPD mutants (Dettmer, U., et al. (2015). Nat. Commun. 6:7314). Clone SOY1 targets a C-terminal half epitope present in all three human spliced iisoforms, NACP140, NACP112, and NACP126, reported by UniProt (P37840). Clone SOY1 also exhibits murine cross-reactivity, albeit at a lower sensitivity than that of human alpha-synuclein.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.