Learn More
SYP Mouse anti-Human, Clone: OTI1C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506521

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.Specifications
SYP | |
Monoclonal | |
Unconjugated | |
SYP | |
MRX96, MRXSYP | |
Mouse | |
Affinity Chromatography | |
RUO | |
6855 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08247 | |
SYP | |
Full length human recombit protein of human SYP produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.