Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYPL1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176316PCP
Description
SYPL1 Polyclonal antibody specifically detects SYPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
SYPL1 | |
Polyclonal | |
Western Blot, ELISA | |
H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL | |
Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SYPL1. The immunogen is located within the first 50 amino acids of SYPL1. Amino Acid Squence: APNIYLVRQRISRLGQ | |
0.1 mL | |
Neuroscience | |
6856 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction