Learn More
Trajan Scientific and Medical™ Syringe
SDP
Catalog No. 000440 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
0.5 μL
Catalog No. Volume (Metric)
00-044-0 0.5 μL
1 options
Catalog No. 00-044-0 Supplier Trajan Scientific and Medical™ Supplier No. 000440
For reduced non specific sample interaction and increased analyte recovery
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Shimadzu instruments
Specifications
Needle Gauge 26 G
Diameter (Metric) Inner Needle 0.155 mm
Diameter (Metric) Outer Needle 0.47 mm
Product Type Syringe
Quantity 1/Ea.
Volume (Metric) 0.5 μL
Needle Point Style Cone
For Use With (Equipment) Shimadzu