Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548935
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548935 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548935 Supplier Invitrogen™ Supplier No. PA548935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYS1 Polyclonal Antibody for Western Blot

Involved in protein trafficking. May serve as a receptor for ARFRP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYS1
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SYS1
Gene Accession No. Q8N2H4
Gene Alias C20orf169; dJ453C12.4; dJ453C12.4.1; Protein SYS1 homolog; SYS1; Sys1 golgi trafficking protein; SYS1 Golgi-localized integral membrane protein homolog
Gene Symbols SYS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 125-153 amino acids from the C-terminal region of human SYS1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.