Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ System Reagents for Phadia™ 1000

Catalog No. 10947601P
Click to view available options
For Use With (Equipment):
Phadia 1000
Phadia™ 1000
Quantity:
∼1200
2 Procedures
2 x 5 L
6 x ∼1200
Product Type:
Assay System Reagent for Phadia 1000
Maintenance solution kit
Stop solution
Washing solution
This item is not returnable. View return policy
This item is not returnable. View return policy

Replacement reagents provide true accuracy, high precision and outstanding quality

TRUSTED_SUSTAINABILITY

Specifications

Product Type Maintenance solution kit
For Use With (Equipment) Phadia™ 1000
Quantity 2 Procedures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.