Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ System Reagents for Phadia™ 1000
Click to view available options
For Use With (Equipment):
Phadia 1000
Phadia™ 1000
Quantity:
∼1200
2 Procedures
2 x 5 L
6 x ∼1200
Product Type:
Assay System Reagent for Phadia 1000
Maintenance solution kit
Stop solution
Washing solution
Specifications
Specifications
Product Type | Stop solution |
For Use With (Equipment) | Phadia 1000 |
Quantity | ∼1200 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction