Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563126 Supplier Invitrogen™ Supplier No. PA563126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSFHKEDGVN SLCQKALHIV TELCFAGQVE WEKCSGIFPR DRGSQGGSST DIS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

SYT10 is a Ca2+ sensor specifically required for the Ca2+-dependent exocytosis of secretory vesicles containing IGF1 in neurons of the olfactory bulb. Exocytosis of IGF1 is required for sensory perception of smell. SYT10 is not involved in Ca2+-dependent synaptic vesicle exocytosis. It acts through Ca2+ and phospholipid binding to the C2 domain: Ca2+ induces binding of the C2-domains to phospholipid membranes and to assembled SNARE-complexes; both actions contribute to triggering exocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYT10
Gene Accession No. Q6XYQ8
Gene Alias DKFZp781D2042; P65; SVP65; synaptotagmin 10; synaptotagmin X; Synaptotagmin-10; SYT; Syt10; SYT-10; SytX
Gene Symbols SYT10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYT10. Recombinant protein control fragment (Product #RP-101200).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.