Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYT11 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298496APC
Description
SYT11 Polyclonal antibody specifically detects SYT11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
SYT11 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
DKFZp781D015, KIAA0080synaptotagmin 12, MGC10881, MGC17226, synaptotagmin XISYT12, synaptotagmin-11, SytXI | |
Produced in rabbits immunized with E. coli-derived Human SYT11 fragment. | |
0.1 mL | |
Neuronal Cell Markers, Neurotransmission | |
23208 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction