Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYT11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SYT11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP169190
SDP
View Documents
Novus Biologicals
NBP169190
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SYT11 Polyclonal specifically detects SYT11 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SYT11
Polyclonal
Rabbit
Neuronal Cell Markers, Neurotransmission
DKFZp781D015, KIAA0080synaptotagmin 12, MGC10881, MGC17226, synaptotagmin XISYT12, synaptotagmin-11, SytXI
SYT11
IgG
48 kDa
Western Blot
Unconjugated
RUO
Q9BT88
23208
Synthetic peptides corresponding to SYT11 (synaptotagmin XI) The peptide sequence was selected from the N terminal of SYT11. Peptide sequence AGLLSRDKDPRGPSSGSCIDQLPIKMDYGEELRSPITSLTPGESKTTSPS.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.