Learn More
SYT11 Mouse anti-Human, Clone: OTI3F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810985

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.Specifications
SYT11 | |
Monoclonal | |
Unconjugated | |
SYT11 | |
SYT12, sytXI | |
Mouse | |
Affinity Chromatography | |
RUO | |
23208 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BT88 | |
SYT11 | |
Human recombit protein fragment corresponding to amino acids 42-296 of human SYT11 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.