Learn More
Invitrogen™ SYT11 Recombinant Rabbit Monoclonal Antibody (2M4R9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555562

Description
Immunogen Sequence: WSCCHQQAEK KQKNPPYKFI HMLKGISIYP ETLSNKKKII KVRRDKDGPG REGGRRNLLV DAAEAGLLSR DKDPRGPSSG SCIDQLPIKM DYGEELRSPI TSLTPGESKT TSPSSPEEDV MLGSLTFSVD YNFPKKALVV TIQEAHGLPV MDDQTQGSDP YIKM.
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature. [provided by RefSeq, Apr 2010].
Specifications
SYT11 | |
Recombinant Monoclonal | |
1.31 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O08835, Q9BT88, Q9R0N3 | |
SYT11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human Synaptotagmin XI (SYT11). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
2M4R9 | |
Unconjugated | |
SYT11 | |
1500004A13Rik; 3632445O20Rik; 5430404N14Rik; 6530420C11Rik; AI851970; KIAA0080; synaptotagmin 11; synaptotagmin 12; synaptotagmin XI; Synaptotagmin-11; SYT11; SYT12; SytXI | |
Rabbit | |
Protein A | |
RUO | |
229521, 23208, 60568 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.