Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYT12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552771 Supplier Invitrogen™ Supplier No. PA552771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLSLSYLPTA ERLTVVVVKA KNLIWTNDKT TADPFVKVYL LQDGRKMSKK KTAVKRDDPN PVFNEAMIFS VPAIVLQDLS LRVTVAESSS DGRGDNVGHV IIGPSASGMG TTHWNQMLAT LRRPVSMWHA V Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. Studies of the orthologous gene in rat have shown that the encoded protein selectively modulates spontaneous synaptic-vesicle exocytosis and may also be involved in regulating calcium independent secretion in nonneuronal cells. Alternative splicing results in multiple transcript variants. The gene has previously been referred to as synaptotagmin XI but has been renamed synaptotagmin XII to be standard with mouse and rat official nomenclature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYT12
Gene Accession No. Q8IV01
Gene Alias Srg1; synaptotagmin 12; synaptotagmin XII; Synaptotagmin12; Synaptotagmin-12; synaptotagmin-related gene 1 protein; SynaptotagminXII; synaptotagmin-XII; Syt XII; SYT11; Syt12; SYT-12; Sytr1; SytXII
Gene Symbols SYT12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYT12. Recombinant protein control fragment (Product #RP-90446).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91683
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.