Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYT12 Polyclonal Antibody
Supplier: Thermo Scientific PA5118298
This item is not returnable.
View return policy
Description

Specifications
SYT12 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q8IV01 | |
SYT12 | |
E. coli-derived Human SYT12 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
SYT12 | |
Srg1; synaptotagmin 12; synaptotagmin XII; Synaptotagmin12; Synaptotagmin-12; synaptotagmin-related gene 1 protein; SynaptotagminXII; synaptotagmin-XII; Syt XII; SYT11; Syt12; SYT-12; Sytr1; SytXII | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
91683 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction