Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561190 Supplier Invitrogen™ Supplier No. PA561190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLTLTLRTCD RFSRHSVAGE LRLGLDGTSV PLGAAQWGEL KTSAKEPSAG AGEVLLSISY LPAANRLLVV LIKAKNLHSN QSKE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.

SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYT13
Gene Accession No. Q7L8C5
Gene Alias 5730409J20Rik; AI549909; Kiaa1427; mKIAA1427; synaptotagmin 13; synaptotagmin XIII; Synaptotagmin-13; SYT13; SytXIII
Gene Symbols SYT13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYT13. Recombinant protein control fragment (Product #RP-95553).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.