Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYT13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111438
Description
Immunogen sequence: KDVSVKVTLK HQARKLKKKQ TKRAKHKINP VWNEMIMFEL PDDLLQASSV E.
SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.
Specifications
| SYT13 | |
| Polyclonal | |
| Unconjugated | |
| SYT13 | |
| 5730409J20Rik; AI549909; Kiaa1427; mKIAA1427; synaptotagmin 13; synaptotagmin XIII; Synaptotagmin-13; SYT13; SytXIII | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L8C5 | |
| SYT13 | |
| Recombinant Protein corresonding to Human SYT13. Recombinant protein control fragment (Product #RP-106716). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction