Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYT16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552072 Supplier Invitrogen™ Supplier No. PA552072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRERMMGEKL FYLSHLHPEG EMKVTLVLEP RSNISSGGSP LSPSAVSHSD STSSTQSLSH GGAPELLVGL SYNATTGRLS VEMIKGSHFR NLAVNRAPDT YGKLFLLNSV GQEMSRCKTS IRRGQPNPVY KETFVFQVAL FQLSDVTLM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

SYT16 is a protein coding gene. May be involved in the trafficking and exocytosis of secretory vesicles in non-neuronal tissues. Is Ca(2+)-independent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYT16
Gene Accession No. Q17RD7
Gene Alias chr14 synaptotagmin; CHR14SYT; CTD-2277K2.1; STREP14; Synaptotagmin 14-like protein; synaptotagmin 16; synaptotagmin XIV-related protein; synaptotagmin XVI; synaptotagmin-16; SYT14L; syt14r; SYT16; yt14r
Gene Symbols SYT16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYT16. Recombinant protein control fragment (Product #RP-88932).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.