Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYT16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101919
Description
Antibody detects endogenous levels of total SYT16.
SYT16 is a protein coding gene. May be involved in the trafficking and exocytosis of secretory vesicles in non-neuronal tissues. Is Ca(2+)-independent.
Specifications
SYT16 | |
Polyclonal | |
Unconjugated | |
SYT16 | |
chr14 synaptotagmin; CHR14SYT; CTD-2277K2.1; STREP14; Synaptotagmin 14-like protein; synaptotagmin 16; synaptotagmin XIV-related protein; synaptotagmin XVI; synaptotagmin-16; SYT14L; syt14r; SYT16; yt14r | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83851 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q17RD7 | |
SYT16 | |
A synthesized peptide derived from human SYT16(Accession Q17RD7), corresponding to amino acid residues L232-H282. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction