Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYT17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114270 Supplier Invitrogen™ Supplier No. PA5114270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLLPDQKNSK QTGVKRKTQK PVFEERYTFE IPFLEAQRRT LLLTVVDFDK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

Plays a role in dendrite formation by melanocytes. Classified as a brain and kidney protein (not to be confused with the BigK K+ channel); may play a role in cAMP mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SYT17
Gene Accession No. Q9BSW7
Gene Alias B/K protein; Bk; brain and kidney protein; protein B/K; synaptotagmin 17; synaptotagmin XVII; Synaptotagmin-17; Syt17; SytXVII
Gene Symbols SYT17
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Human SYT17.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51760
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.