Learn More
SYT4 Mouse anti-Human, Clone: OTI3B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505864

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in Ca(2+)-dependent exocytosis of secretory vesicles through Ca(2+) and phospholipid binding to the C2 domain or may serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis.Specifications
SYT4 | |
Monoclonal | |
Unconjugated | |
SYT4 | |
HsT1192 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6860 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H2B2 | |
SYT4 | |
Full length human recombit protein of human SYT4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.