Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syt7 Mouse anti-Human, Mouse, Rat, APC, Clone:S275-14, Abnova™

Catalog No. 89333846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-846 Supplier Abnova Corporation Supplier No. MAB17036
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant mouse Syt7.

Specifications

Antigen Syt7
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S275-14
Conjugate APC
Description synaptotagmin VII
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Syt7
Gene Alias AI851541, B230112P13Rik
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 150-239 at cytoplasmic C2A domain of mouse Syt7.
Quantity 100 μg
Gene ID (Entrez) 54525
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.