Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558126 Supplier Invitrogen™ Supplier No. PA558126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAIAGVLQRD ARLRQLEEGR VSKLRASVAD PGQLKILTGD WFQEARSQRH HNAHFGSDLV RASMRRKKST RGDQAPGHDR EAEAAVKEK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

SYTL1 may play a role in vesicle trafficking. Binds phosphatidylinositol 3,4,5-trisphosphate. Acts as a RAB27A effector protein and may play a role in cytotoxic granule exocytosis in lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYTL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYTL1
Gene Accession No. Q8IYJ3
Gene Alias exophilin-7; FLJ14996; JFC1; NADPH oxidase-related, C2 domain-containing protein; Protein JFC1; Protein SLP1; SB146; SLP1; synaptotagmin like 1; synaptotagmin-like 1; synaptotagmin-like protein 1; SYTL1
Gene Symbols SYTL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYTL1. Recombinant protein control fragment (Product #RP-94260).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.