Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114163 Supplier Invitrogen™ Supplier No. PA5114163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAEAAVKEKE EGPEPRLTID EAPQERLRET EASDPEEASQ AQEDPGQGDQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

SYTL1 may play a role in vesicle trafficking. Binds phosphatidylinositol 3,4,5-trisphosphate. Acts as a RAB27A effector protein and may play a role in cytotoxic granule exocytosis in lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYTL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SYTL1
Gene Accession No. Q8IYJ3
Gene Alias exophilin-7; FLJ14996; JFC1; NADPH oxidase-related, C2 domain-containing protein; Protein JFC1; Protein SLP1; SB146; SLP1; synaptotagmin like 1; synaptotagmin-like 1; synaptotagmin-like protein 1; SYTL1
Gene Symbols SYTL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SYTL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84958
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.