Learn More
Invitrogen™ SYTL2 Polyclonal Antibody

Description
SYTL2 Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | SYTL2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SYTL2 |
| Gene Accession No. | Q9HCH5 |
| Gene Alias | AI266830; Breast cancer-associated antigen SGA-72M; CHR11SYT; chromosome 11 synaptotagmin; EXO4; Exophilin-4; KIAA1597; mKIAA1597; PPP1R151; protein phosphatase 1, regulatory subunit 151; SGA72M; SLP2; SLP2A; synaptotagmin like 2; synaptotagmin-like 2; synaptotagmin-like protein 2; SYTL2 |
| Gene Symbols | SYTL2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.