Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556513 Supplier Invitrogen™ Supplier No. PA556513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THAWKCTVCF EDRNVKIKTG EWFYEERAKK FPTGGKHETV GGQLLQSYQK LSKISVVPPT PPPVSESQCS RSPGRLQEFG QFRGFNKSVE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

May act as Rab effector protein and play a role in vesicle trafficking. Binds phospholipids in the presence of calcium ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYTL3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYTL3
Gene Accession No. Q4VX76
Gene Alias A430092N21; Exophilin-6; mFLJ00307; RGD1561992; Slp3; Slp3-a; Slp3-b; synaptotagmin like 3; synaptotagmin-like 3; synaptotagmin-like protein 3; Sytl3
Gene Symbols SYTL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYTL3. Recombinant protein control fragment (Product #RP-94936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94120
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.