Learn More
Invitrogen™ SYTL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551585
Description
Immunogen sequence: GKSALEAESE SLDSFTADSD STSRRDSLDK SGLFPEWKKM SAPKSQVEKE TQPGGQNVVF VDEGEMIFKK NTRKILRPSE YTKSVIDLRP EDVVHESGSL GDRSKSVPGL NVDMEEEEEE EDIDHLVKLH RQKLARSSMQ SGSSM Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
SYTL4 | |
Polyclonal | |
Unconjugated | |
SYTL4 | |
Exophilin-2; Gph; Granuphilin; granuphilin b; granuphilin-a; granuphilin-b; Slp4; synaptotagmin like 4; synaptotagmin-like 4; synaptotagmin-like 4 (granuphilin-a); synaptotagmin-like protein 4; Sytl4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96C24 | |
SYTL4 | |
Recombinant protein corresponding to Human SYTL4. Recombinant protein control fragment (Product #RP-92123). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.