Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555284 Supplier Invitrogen™ Supplier No. PA555284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQVNVLGTDV VRQSILRRSP AEEVQSQEQT RQDAEKSDTS PVAGKKASHD GPKRKGFLLS KFRSATRGEI ITPKTDTGRS YSLDLDGQHF RSLKSPPGSD RGSTGSSDLN DQEP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.

SYTL5 belongs to the synaptotagmin-like (Slp) protein family, which contains a unique Slp homolog domain (SHD) at the N-terminus. The SHD functions as a binding site for Rab27A, which plays a role in protein transport. Expression of SYTL5 is restricted to placenta and liver, suggesting that it might be involved in Rab27A-dependent membrane trafficking in specific tissues. Several members of the synaptotagmin family are calcium-sensing vesicle-fusion proteins as mediators of cell migration. SYTL7 & SYTL5 were positive regulators of chemotaxis, whereas SYTL2 was a negative regulator of chemotaxis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYTL5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYTL5
Gene Accession No. Q8TDW5
Gene Alias ENSMUSG00000054453; exophilin 9; Slp5; synaptotagmin 15; synaptotagmin like 5; synaptotagmin-like 5; synaptotagmin-like protein 5; Sytl5
Gene Symbols SYTL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYTL5. Recombinant protein control fragment (Product #RP-91897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.