Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTOX™ Green Ready Flow™ Reagent

Catalog No. R37168
Encompass_Preferred
Change view
Click to view available options
Quantity:
120 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37168 120 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37168 Supplier Invitrogen™ Supplier No. R37168
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SYTOX™ Green Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays.

  • A bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays
  • It is excitable at 488 nm with an emission profile similar to FITC, and has been tested in multiple cell lines
  • Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze
  • As a cell-impermeant dye, SYTOX Green enters the cell upon loss of membrane integrity and binds to DNA, thereby acting as a counterstain that can be analyzed when excited
  • SYTOX Green reagent can be used in both mammalian and bacterial cells
  • It is often included in multiplex assays
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Green
Content And Storage Includes 2 dropper bottles, store at room temperature.
Description SYTOX™ Green Ready Flow™ Reagent
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Reagent
Dye Type SYTOX™ Green Ready Flow™
Form Solution
Format Bottle(s)
Product Line Ready Flow, Sytox
Quantity 120 Assays
Solubility DMSO (Dimethylsulfoxide)
Excitation Wavelength 504 nm
Emission Wavelength 523
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.