Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T-2 Mycotoxin Monoclonal Antibody (4A6)

Catalog No. PIMA549615
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549615 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549615 Supplier Invitrogen™ Supplier No. MA549615

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

T-2 mycotoxin is a toxic secondary metabolite produced by several species of fungi, including Fusarium sporotrichioides and Fusarium poae. These fungi can grow on a variety of crops, including wheat, barley, oats, and corn, and can produce T-2 mycotoxin under certain conditions. T-2 mycotoxin is a potent inhibitor of protein synthesis and can cause a variety of toxic effects in animals, including vomiting, diarrhea, hemorrhaging, and immunosuppression. It is also a potential human carcinogen and has been linked to esophageal cancer in some populations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-2 Mycotoxin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4A6
Concentration 4.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Mycotoxin T2; Mycotoxin T-2; T2 Toxin; T-2 Toxin
Host Species Mouse
Immunogen Conjugate of T-2 hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.